Skip to Content
Merck
DL-2-Amino-3-phosphonopropionic acid

DL-2-Amino-3-phosphonopropionic acid

DL-2-Amino-3-phosphonopropionic acid
Synonym(s):
3-Phosphono-DL-alanine, AP-3
Empirical Formula (Hill Notation):
C3H8NO5P
CAS Number:
Molecular Weight:
169.07

Products

Shipping
Product Category
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing