![Dimethyl-d6-amine hydrochloride 99 atom % D](/deepweb/assets/sigmaaldrich/product/structures/152/601/661003ce-cf7f-494d-9f69-97afc025dc83/640/661003ce-cf7f-494d-9f69-97afc025dc83.png)
Dimethyl-d6-amine hydrochloride
![Dimethyl-d6-amine hydrochloride 99 atom % D](/deepweb/assets/sigmaaldrich/product/structures/152/601/661003ce-cf7f-494d-9f69-97afc025dc83/640/661003ce-cf7f-494d-9f69-97afc025dc83.png)
Synonym(s):
Deuterated dimethylamine hydrochloride
Linear Formula:
(CD3)2NH · HCl
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing