Skip to Content
Merck
Diethyl ethylmalonate 99%

Diethyl ethylmalonate

Diethyl ethylmalonate 99%
Synonym(s):
Diethyl ethylmalonate
Linear Formula:
C2H5CH(COOC2H5)2
CAS Number:
Molecular Weight:
188.22

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing