Skip to Content
Merck
(−)-Diethyl D-tartrate ≥99%

(−)-Diethyl D-tartrate

(−)-Diethyl D-tartrate ≥99%
Synonym(s):
D-(−)-Tartaric acid diethyl ester
Linear Formula:
HO2CCCH(OH)CH(OH)CO2H
CAS Number:
Molecular Weight:
206.19
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing