Skip to Content
Merck
Diethyl azodicarboxylate solution purum, ~40% in toluene (H-NMR)

Diethyl azodicarboxylate solution

Diethyl azodicarboxylate solution purum, ~40% in toluene (H-NMR)
Synonym(s):
1,2-Ethoxycarbonyl diazene solution, DEAD, Diethoxycarbonyldiazene solution, Diethyl azodiformate solution, NSC 3474, NSC 679015, Unifoam AZ-AE 200
Linear Formula:
C2H5OCON=NCOOC2H5
CAS Number:
Molecular Weight:
174.15

Products

Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing