Skip to Content
Merck
Dibucaine Hydrochloride certified reference material, pharmaceutical secondary standard

Dibucaine hydrochloride

Dibucaine Hydrochloride certified reference material, pharmaceutical secondary standard
Synonym(s):
2-Butoxy-N-(2-diethylaminoethyl)-4-quinolinecarboxamide hydrochloride, Cinchocaine hydrochloride, Dibucaine hydrochloride
Empirical Formula (Hill Notation):
C20H29N3O2 · HCl
CAS Number:
Molecular Weight:
379.92

Products

Shipping
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique