Skip to Content
Merck
Dibenzothiophene analytical standard

Dibenzothiophene

Dibenzothiophene analytical standard
Synonym(s):
DBT, Diphenylene sulfide, Dibenzothiophene
Empirical Formula (Hill Notation):
C12H8S
CAS Number:
Molecular Weight:
184.26
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing