![[D-Arg1, D-Phe5, D-Trp7,9, Leu11]-Substance P ≥95% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/868/763/c158a0f4-4ad9-4d15-b940-1dd6a7f30697/640/c158a0f4-4ad9-4d15-b940-1dd6a7f30697.png)
[D-Arg1, D-Phe5, D-Trp7,9, Leu11]-Substance P
![[D-Arg1, D-Phe5, D-Trp7,9, Leu11]-Substance P ≥95% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/868/763/c158a0f4-4ad9-4d15-b940-1dd6a7f30697/640/c158a0f4-4ad9-4d15-b940-1dd6a7f30697.png)
Synonym(s):
D-Arg-Pro-Lys-Pro-D-Phe-Gln-D-Trp-Phe-D-Trp-Leu-Leu-NH2, Antagonist D
Empirical Formula (Hill Notation):
C79H109N19O12
CAS Number:
Molecular Weight:
1516.83
Products
Product Category
Brand
Formula Weight
Gene Alias
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing