Dansylhydrazine
Synonym(s):
5-(Dimethylamino)-1-naphthalenesulfonic hydrazide, 5-(Dimethylamino)naphthalene-1-sulfonohydrazide, Dns-Hz
Linear Formula:
(CH3)2NC10H6SO2NHNH2
Products
Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing