![(+)-3-Methoxymorphinan hydrochloride analytical standard, for drug analysis](/deepweb/assets/sigmaaldrich/product/structures/183/867/a46f14cb-61e9-47b2-b8b6-5e2cf0a09ab6/640/a46f14cb-61e9-47b2-b8b6-5e2cf0a09ab6.png)
(+)-3-Methoxymorphinan hydrochloride
![(+)-3-Methoxymorphinan hydrochloride analytical standard, for drug analysis](/deepweb/assets/sigmaaldrich/product/structures/183/867/a46f14cb-61e9-47b2-b8b6-5e2cf0a09ab6/640/a46f14cb-61e9-47b2-b8b6-5e2cf0a09ab6.png)
Synonym(s):
(+)-cis-1,3,4,9,10,10a-Hexahydro-6-methoxy-2H-10,4a-iminoethanophenanthrene
Empirical Formula (Hill Notation):
C17H23NO · HCl
CAS Number:
Molecular Weight:
293.83
Products
Brand
Color
Formula Weight
Physical Form
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing