CyJohnPhos
Synonym(s):
2-(Dicyclohexylphosphino)biphenyl, Cyclohexyl JohnPhos
Empirical Formula (Hill Notation):
C24H31P
CAS Number:
Molecular Weight:
350.48
Products
Shipping
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing