Skip to Content
Merck
Cyclopentadienyl iron(II) dicarbonyl dimer 99%

Cyclopentadienyl iron(II) dicarbonyl dimer

Cyclopentadienyl iron(II) dicarbonyl dimer 99%
Synonym(s):
Bis(cyclopentadienyl)tetracarbonyl-diiron, Di(cyclopentadienyl)tetracarbonyl-diiron
Empirical Formula (Hill Notation):
C14H10Fe2O4
CAS Number:
Molecular Weight:
353.92
EC Number:

Products

Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing