Cycloartenol
Synonym(s):
(3S,5R,8S,9S,10R,13R,14S,17R)-17-((R)-1,5-Dimethyl-hex-4-enyl)-4,4,13,14-tetramethyl-tetradecahydro-cyclopropa[9,10]cyclopenta[a]phenanthren-3-ol, 9,19-Cyclo-24-lanosten-3β-ol, Handianol
Empirical Formula (Hill Notation):
C30H50O
CAS Number:
Molecular Weight:
426.72
Products
Shipping
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing