Cryptotanshinone
Synonym(s):
1,2,6,7,8,9-hexahydro-1,6,6-trimethyl- (R)-phenanthro(1,2-b)furan-10,11-dione, Cryptotanshinon, Tanshinone c, (R)-(−)-1,6,6-Trimethyl-1,2,6,7,8,9-hexahydrophenanthro[1,2-b]furan-10,11-dione, Tanshinone C
Empirical Formula (Hill Notation):
C19H20O3
CAS Number:
Molecular Weight:
296.36
Products
Shipping
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing