Skip to Content
Merck
Coumarin 153 Dye content 99 %

Coumarin 153

Coumarin 153 Dye content 99 %
Synonym(s):
2,3,6,7-Tetrahydro-9-(trifluoromethyl)-1H,5H,11H-[1]benzopyrano(6,7,8-ij)quinolizin-11-one, 2,3,6,7-Tetrahydro-9-trifluoromethyl-1H,5H-quinolizino(9,1-gh)coumarin, 8-Trifluoromethyl-2,3,5,6-4H-1,H-11-oxa-3a-aza-benzo[de]anthracen-10-one, Coumarin 540A
Empirical Formula (Hill Notation):
C16H14F3NO2
CAS Number:
Molecular Weight:
309.28
EC Number:

Products

Shipping
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing