Skip to Content
Merck
Copper(II) nitrate hydrate ≥99.99% trace metals basis

Copper(II) nitrate hydrate

Copper(II) nitrate hydrate ≥99.99% trace metals basis
Synonym(s):
Cupric nitrate hydrate
Linear Formula:
Cu(NO3)2 · xH2O
CAS Number:
Molecular Weight:
187.56 (anhydrous basis)

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group