Skip to Content
Merck
Copper(II) acetate monohydrate for analysis EMSURE® ACS

Copper(II) acetate monohydrate

Copper(II) acetate monohydrate for analysis EMSURE® ACS
Synonym(s):
Copper(II) acetate monohydrate
Linear Formula:
(CH3COO)2Cu · H2O
CAS Number:
Molecular Weight:
181.63 (anhydrous basis)

Products

Product Category
Brand
Sustainability
Feature
Formula Weight
Physical Form
Product Line
Quality Segment
Showing 1-2 of 2