
Clopidogrel Related Compound B

Synonym(s):
(RS)-Methyl (2-chlorophenyl)-4,5-dihydrothieno[2,3-c]pyridine-6(7H)-acetate hydrochloride
Empirical Formula (Hill Notation):
C16H16ClNO2S · HCl
CAS Number:
Molecular Weight:
358.28
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing