Skip to Content
Merck
Clindamycin 2-phosphate aminoglycoside antibiotic

Clindamycin 2-phosphate

Clindamycin 2-phosphate aminoglycoside antibiotic
Synonym(s):
Clindamycin 2-dihydrogen phosphate
Empirical Formula (Hill Notation):
C18H34ClN2O8PS
CAS Number:
Molecular Weight:
504.96
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Activity Spectrum
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing