Skip to Content
Merck
Clarithromycin impurity E British Pharmacopoeia (BP) Reference Standard

Clarithromycin impurity E

Clarithromycin impurity E British Pharmacopoeia (BP) Reference Standard
Synonym(s):
6,11-Di-O-methylerythromycin A
Empirical Formula (Hill Notation):
C39H71NO13
CAS Number:
Molecular Weight:
761.98

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing