Citral
Synonym(s):
3,7-Dimethyl-2,6-octadienal, Geranial and neral mixture, Citral
Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCHO
CAS Number:
Molecular Weight:
152.23
Products
Shipping
Product Category
Brand
Biological Source
Boiling Point (°C)
Color
Feature
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Chemical Composition
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing