![Ciprofloxacin hydrochloride British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/374/908/7a74a36e-fb89-4748-9c8d-92508e177129/640/7a74a36e-fb89-4748-9c8d-92508e177129.png)
Ciprofloxacin hydrochloride
![Ciprofloxacin hydrochloride British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/374/908/7a74a36e-fb89-4748-9c8d-92508e177129/640/7a74a36e-fb89-4748-9c8d-92508e177129.png)
Synonym(s):
Ciprofloxacin hydrochloride monohydrate
Empirical Formula (Hill Notation):
C17H18FN3O3 · HCl · H2O
CAS Number:
Molecular Weight:
385.82
Products
Shipping
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3