Skip to Content
Merck
Cinacalcet hydrochloride ≥98% (HPLC)

Cinacalcet hydrochloride

Cinacalcet hydrochloride ≥98% (HPLC)
Synonym(s):
AMG 073 hydrochloride, KRN 1493 hydrochloride, N-[1-(R)-(-)-(1-naphthyl)ethyl]-3-[3(trifluoromethyl)phenyl]-1-aminopropane hydrochloride, NPS 1493 hydrochloride
Empirical Formula (Hill Notation):
C22H22F3N·HCl
CAS Number:
Molecular Weight:
393.87

Products

Shipping
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2