Skip to Content
Merck
Chlorosuccinic acid 96%

Chlorosuccinic acid

Chlorosuccinic acid 96%
Synonym(s):
2-Chlorobutanedioic acid
Linear Formula:
HOOCCH2CHClCOOH
CAS Number:
Molecular Weight:
152.53

Products

Shipping
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing