Skip to Content
Merck
(Chloromethyl)trimethylsilane 98%

(Chloromethyl)trimethylsilane

(Chloromethyl)trimethylsilane 98%
Synonym(s):
(Trimethylsilyl)methyl chloride, (Chloromethyl)trimethylsilane
Linear Formula:
(CH3)3SiCH2Cl
CAS Number:
Molecular Weight:
122.67

Products

Shipping
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing