Chicoric acid
Synonym(s):
(−)-Chicoric acid, (2R,3R)-2,3-Bis{[(2E)-3-(3,4-dihydroxyphenyl)-1-oxo-2-propen-1-yl]oxy}butanedioic acid, 2,3-Di-trans-caffeoyltartaric acid
Empirical Formula (Hill Notation):
C22H18O12
CAS Number:
Molecular Weight:
474.37
Products
Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing