Skip to Content
Merck
Cetrimonium bromide United States Pharmacopeia (USP) Reference Standard

Cetrimonium bromide

Cetrimonium bromide United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Hexadecyltrimethylammonium bromide, CTAB, Cetrimonium bromide, Cetyltrimethylammonium bromide, Palmityltrimethylammonium bromide
Linear Formula:
CH3(CH2)15N(Br)(CH3)3
CAS Number:
Molecular Weight:
364.45

Products

Shipping
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing