Skip to Content
Merck
Cellulose acetate propionate average Mn ~15,000 by GPC, powder

Cellulose acetate propionate

Cellulose acetate propionate average Mn ~15,000 by GPC, powder
Empirical Formula (Hill Notation):
C76H114O49
CAS Number:
Molecular Weight:
1811.69

Products

Shipping
Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Polymer Composition
Polymer Type