Skip to Content
Merck
Cellotetraose ≥85% (HPLC)

Cellotetraose

Cellotetraose ≥85% (HPLC)
Synonym(s):
(β-D-Glc-[1→4])3-D-Glc, D-(+)-Cellotetraose
Empirical Formula (Hill Notation):
C24H42O21
CAS Number:
Molecular Weight:
666.58

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing