Skip to Content
Merck
Ceftriaxone disodium salt hemi(heptahydrate) third-generation cephalosporin antibiotic

Ceftriaxone disodium salt hemi(heptahydrate)

Ceftriaxone disodium salt hemi(heptahydrate) third-generation cephalosporin antibiotic
Empirical Formula (Hill Notation):
C18H16N8Na2O7S3 · 3.5H2O
CAS Number:
Molecular Weight:
661.60
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Activity Spectrum
Mode of Action
Showing 1-1 of 1