Skip to Content
Merck
cataCXium A Pd G4

cataCXium A Pd G4

cataCXium A Pd G4
Empirical Formula (Hill Notation):
C38H54NO3PPdS
CAS Number:
Molecular Weight:
742.30

Products

Shipping
Brand
Formula Weight
Physical Form
Quality Segment
Functional Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing