Skip to Content
Merck
vibrant-m

Carmustine impurity A

vibrant-m
Synonym(s):
1,3-Bis(2-chloroethyl)urea, BCU, NSC 36198
Empirical Formula (Hill Notation):
C5H10Cl2N2O
CAS Number:
Molecular Weight:
185.05

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing