Canrenone
Synonym(s):
(17α)-17-Hydroxy-3-oxopregna-4,6-diene-21-carboxylic acid γ-lactone, NSC 261713
Empirical Formula (Hill Notation):
C22H28O3
CAS Number:
Molecular Weight:
340.46
Products
Shipping
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing