![Cannabinol solution 1 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/284/471/1d69a4cc-f7ff-425c-b03e-8474976f9a40/640/1d69a4cc-f7ff-425c-b03e-8474976f9a40.png)
Cannabinol solution
![Cannabinol solution 1 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/284/471/1d69a4cc-f7ff-425c-b03e-8474976f9a40/640/1d69a4cc-f7ff-425c-b03e-8474976f9a40.png)
Empirical Formula (Hill Notation):
C21H26O2
CAS Number:
Molecular Weight:
310.43
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2