Skip to Content
Merck
(−)-Camphor-10-sulfonic acid purum, ≥98.0% (T)

(−)-Camphor-10-sulfonic acid

(−)-Camphor-10-sulfonic acid purum, ≥98.0% (T)
Synonym(s):
(1R)-(−)-Camphor-10-sulfonic acid
Empirical Formula (Hill Notation):
C10H16O4S
CAS Number:
Molecular Weight:
232.30
EC Number:

Products

Shipping
Product Category
Brand
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing