Skip to Content
Merck
Calcium ionophore II Selectophore™, function tested

Calcium ionophore II

Calcium ionophore II Selectophore™, function tested
Synonym(s):
N,N,N′,N′-Tetra[cyclohexyl]diglycolic acid diamide, N,N,N′,N′-Tetracyclohexyl-3-oxapentanediamide, ETH 129
Empirical Formula (Hill Notation):
C28H48N2O3
CAS Number:
Molecular Weight:
460.69

Products

Shipping
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing