Skip to Content
Merck
Calcium gluconate monohydrate United States Pharmacopeia (USP) Reference Standard

Calcium gluconate monohydrate

Calcium gluconate monohydrate United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Calcium D-gluconate monohydrate, D-Gluconic acid calcium salt
Empirical Formula (Hill Notation):
C12H22CaO14 · H2O
CAS Number:
Molecular Weight:
448.39

Products

Shipping
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1