Skip to Content
Merck
Butyric anhydride purum, ≥97.0% (NT)

Butyric anhydride

Butyric anhydride purum, ≥97.0% (NT)
Synonym(s):
Butanoic anhydride, Butyric anhydride
Linear Formula:
(CH3CH2CH2CO)2O
CAS Number:
Molecular Weight:
158.19
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing