Skip to Content
Merck
Buffered Peptone Water ReadyBag®, ISO, FDA BAM, EP, granular, pack of 35 × 86 g, sterile; γ-irradiated

Buffered Peptone Water

Buffered Peptone Water ReadyBag®, ISO, FDA BAM, EP, granular, pack of 35 × 86 g, sterile; γ-irradiated

Products

Shipping
Product Category
Brand
Color
pH Value
Physical Form
Agency
Chemical Composition
Capacity (mL)
Quality Segment
Closure Type
Application
Microorganism Suitability
Package Size
Sterility
Sterilization Method
Technique
Showing 1-11 of 11
Compare
Product Number
Description
Pricing