
Brucine

Synonym(s):
10,11-Dimethoxystrychnine, 2,3-Dimethoxystrychnidin-10-one
Empirical Formula (Hill Notation):
C23H26N2O4
CAS Number:
Molecular Weight:
394.46
Products
Shipping
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing