Skip to Content
Merck
Bromobimane ≥97% (HPLC)

Bromobimane

Bromobimane ≥97% (HPLC)
Synonym(s):
Monobromobimane
Empirical Formula (Hill Notation):
C10H11BrN2O2
CAS Number:
Molecular Weight:
271.11

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing