Skip to Content
Merck
Bromadiolon PESTANAL®, analytical standard

Bromadiolon

Bromadiolon PESTANAL®, analytical standard
Synonym(s):
Bromodiolone
Empirical Formula (Hill Notation):
C30H23BrO4
CAS Number:
Molecular Weight:
527.41
EC Number:

Products

Shipping
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing