Skip to Content
Merck
Boc-Phe-OH ≥99.0% (T)

Boc-Phe-OH

Boc-Phe-OH ≥99.0% (T)
Synonym(s):
N-(tert-Butoxycarbonyl)-L-phenylalanine, Boc-L-phenylalanine, Boc-Phe-OH
Linear Formula:
C6H5CH2CH(COOH)NHCOOC(CH3)3
CAS Number:
Molecular Weight:
265.30

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Reagent Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing