Skip to Content
Merck
Bis(triphenylphosphoranylidene)ammonium chloride 97%

Bis(triphenylphosphoranylidene)ammonium chloride

Bis(triphenylphosphoranylidene)ammonium chloride 97%
Synonym(s):
Bis(triphenylphosphine)iminium chloride, Hexaphenyldiphosphazenium chloride, PPNCl
Linear Formula:
(C6H5)3P=N(Cl)=P(C6H5)3
CAS Number:
Molecular Weight:
574.03
EC Number:

Products

Shipping
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing