
Bisphenol A bis(3-chloro-2-hydroxypropyl) ether

Synonym(s):
2,2-Bis[4-(3-chloro-2-hydroxypropoxy)phenyl]propane, BADGE-2HCl
Empirical Formula (Hill Notation):
C21H26Cl2O4
CAS Number:
Molecular Weight:
413.33
Products
Shipping
Formula Weight
Purity
Format
Quality Segment
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing