![β-Ionone natural, ≥95%, FG](/deepweb/assets/sigmaaldrich/product/structures/381/632/d72807e1-8fb6-4283-9c68-de6e775b5b2a/640/d72807e1-8fb6-4283-9c68-de6e775b5b2a.png)
β-Ionone
![β-Ionone natural, ≥95%, FG](/deepweb/assets/sigmaaldrich/product/structures/381/632/d72807e1-8fb6-4283-9c68-de6e775b5b2a/640/d72807e1-8fb6-4283-9c68-de6e775b5b2a.png)
Synonym(s):
4-(2,6,6-Trimethyl-1-cyclohexenyl)-3-buten-2-one
Empirical Formula (Hill Notation):
C13H20O
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing