Bifonazole
Synonym(s):
1-(p,α-Diphenylbenzyl)imidazole
Empirical Formula (Hill Notation):
C22H18N2
CAS Number:
Molecular Weight:
310.39
Products
Shipping
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3