Skip to Content
Merck
Benzene-D6 ≥95 atom % D, ≥99% (CP)

Benzene-D6

Benzene-D6 ≥95 atom % D, ≥99% (CP)
Synonym(s):
Hexadeuterobenzene, Benzene-D6
Empirical Formula (Hill Notation):
C6D6
CAS Number:
Molecular Weight:
84.15

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Application
Special Grade
Technique