Skip to Content
Merck
Benzalkonium Chloride 10% Solution Pharmaceutical Secondary Standard; Certified Reference Material

Benzalkonium chloride solution

Benzalkonium Chloride 10% Solution Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Alkylbenzyldimethylammonium chloride, Benzalkonium chloride solution
CAS Number:

Products

Shipping
Product Category
Brand
Biological Source
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade