
Barium perchlorate

Synonym(s):
Barium diperchlorate, Perchloric acid barium salt, Barium perchlorate
Linear Formula:
Ba(ClO4)2
CAS Number:
Molecular Weight:
336.23
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Reagent Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing